Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pxylp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079153.1 | NP_001072621.1 | 5 - 6 | 523 | 79 | |
XM_012970560.2 | XP_012826014.1 | 5 - 6 | 572 | 79 | ||
GenBank mRNA | BC123016.1 | - | 5 - 6 | 523 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |