Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh2d4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079236.2 | NP_001072704.1 | 3 - 4 | 673 | 115 | |
XM_012965810.2 | XP_012821264.1 | 4 - 5 | 810 | 115 | ||
XM_012965811.2 | XP_012821265.1 | 5 - 6 | 913 | 115 | ||
XM_012965812.2 | XP_012821266.1 | 4 - 5 | 760 | 115 | ||
GenBank mRNA | BC124048.1 | - | 3 - 4 | 674 | 115 | |
CR760968.2 | - | 4 - 5 | 735 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |