Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paqr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079240.1 | NP_001072708.1 | 4 - 5 | 472 | 80 | |
XM_012959271.2 | XP_012814725.1 | 5 - 6 | 751 | 80 | ||
XM_012959273.2 | XP_012814727.1 | 4 - 5 | 542 | 80 | ||
XM_012959274.2 | XP_012814728.1 | 4 - 5 | 567 | 80 | ||
XM_012959275.2 | XP_012814729.1 | 4 - 5 | 563 | 80 | ||
XM_018092178.1 | XP_017947667.1 | 5 - 6 | 747 | 80 | ||
XM_018092179.1 | XP_017947668.1 | 5 - 6 | 749 | 80 | ||
GenBank mRNA | BC124054.1 | - | 4 - 5 | 472 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Xenopus tropicalis Probe spans exons |