Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paqr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079240.1 | NP_001072708.1 | 5 - 6 | 596 | 77 | |
XM_012959271.2 | XP_012814725.1 | 6 - 7 | 875 | 77 | ||
XM_012959273.2 | XP_012814727.1 | 5 - 6 | 666 | 77 | ||
XM_012959274.2 | XP_012814728.1 | 5 - 6 | 691 | 77 | ||
XM_012959275.2 | XP_012814729.1 | 5 - 6 | 687 | 77 | ||
XM_018092178.1 | XP_017947667.1 | 6 - 7 | 871 | 77 | ||
GenBank mRNA | BC124054.1 | - | 5 - 6 | 596 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Xenopus tropicalis Probe spans exons |