Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079245.1 | NP_001072713.1 | 985 | 72 | ||
GenBank mRNA | BC125667.1 | - | 985 | 72 | ||
CT030525.1 | - | 6 - 7 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |