Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnaset2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079248.1 | NP_001072716.1 | 2 - 3 | 237 | 102 | |
GenBank mRNA | BC125675.1 | - | 2 - 3 | 237 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |