Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnaset2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079248.1 | NP_001072716.1 | 5 - 6 | 425 | 92 | |
GenBank mRNA | BC125675.1 | - | 5 - 6 | 425 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |