Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: htra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079262.2 | NP_001072730.2 | 2 | 724 | 97 | |
GenBank mRNA | BC125705.1 | - | 2 | 724 | 97 | |
BC135434.1 | - | 2 | 647 | 97 | ||
CT025323.2 | - | 2 | 631 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Both primers and probe map within a single exon |