Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: calb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079343.1 | NP_001072811.1 | 9 - 10 | 666 | 98 | |
XM_012964953.1 | XP_012820407.1 | 9 - 10 | 675 | 98 | ||
XM_012964954.1 | XP_012820408.1 | 8 - 9 | 593 | 98 | ||
GenBank mRNA | BC121487.1 | - | 9 - 10 | 666 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |