Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079392.1 | NP_001072860.1 | 4 - 5 | 866 | 97 | |
XM_012970576.2 | XP_012826030.1 | 5 - 6 | 1015 | 97 | ||
GenBank mRNA | BC123066.1 | - | 4 - 5 | 866 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |