Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016053.3 | NP_001016053.2 | 4 - 5 | 327 | 83 | |
GenBank mRNA | BC121508.1 | - | 4 - 5 | 327 | 83 | |
CR762335.2 | - | 4 - 5 | 388 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |