Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016079.3 | NP_001016079.1 | 2 - 3 | 319 | 88 | |
XM_012958991.2 | XP_012814445.1 | 2 - 3 | 391 | 88 | ||
XM_018092069.1 | XP_017947558.1 | 2 - 3 | 405 | 88 | ||
GenBank mRNA | BC121999.1 | - | 2 - 3 | 319 | 88 | |
CR762219.2 | - | 2 - 3 | 239 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |