Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016125.3 | NP_001016125.1 | 5 - 6 | 384 | 87 | |
GenBank mRNA | BC128617.1 | - | 5 - 6 | 384 | 87 | |
CR762066.4 | - | 5 - 6 | 397 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |