Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016125.3 | NP_001016125.1 | 6 - 7 | 443 | 72 | |
GenBank mRNA | BC128617.1 | - | 6 - 7 | 443 | 72 | |
CR762066.4 | - | 6 - 7 | 456 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |