Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016158.3 | NP_001016158.2 | 5 - 6 | 1456 | 112 | |
XM_012968369.2 | XP_012823823.1 | 5 - 6 | 1325 | 112 | ||
GenBank mRNA | BC135714.1 | - | 5 - 6 | 1456 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |