Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tma16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016631.3 | NP_001016631.1 | 4 - 5 | 348 | 99 | |
XM_012962584.2 | XP_012818038.1 | 5 - 6 | 663 | 99 | ||
GenBank mRNA | BC135799.1 | - | 4 - 5 | 348 | 99 | |
CR760447.2 | - | 4 - 5 | 255 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |