Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cox5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017049.3 | NP_001017049.1 | 1 - 2 | 195 | 104 | |
GenBank mRNA | BC135615.1 | - | 1 - 2 | 195 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |