Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017068.3 | NP_001017068.1 | 4 - 5 | 505 | 79 | |
XM_012957025.2 | XP_012812479.1 | 4 - 5 | 618 | 79 | ||
GenBank mRNA | BC135652.1 | - | 4 - 5 | 505 | 79 | |
CR761572.2 | - | 4 - 5 | 495 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |