Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: opcml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079019.1 | NP_001072487.1 | 6 - 7 | 844 | 93 | |
XM_012966745.2 | XP_012822199.1 | 6 - 7 | 1025 | 93 | ||
GenBank mRNA | BC121914.1 | - | 6 - 7 | 844 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |