Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rims3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079023.1 | NP_001072491.1 | 3 - 4 | 395 | 91 | |
GenBank mRNA | BC121920.1 | - | 3 - 4 | 395 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |