Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: guca1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079039.1 | NP_001072507.1 | 2 - 3 | 827 | 75 | |
GenBank mRNA | BC121947.1 | - | 2 - 3 | 827 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |