Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tubg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079041.1 | NP_001072509.1 | 1094 | 71 | ||
GenBank mRNA | BC121949.1 | - | 1094 | 71 |
Gene Symbol: LOC105945133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018097865.1 | XP_017953354.1 | 8 - 9 | 1042 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |