Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079062.1 | NP_001072530.1 | 3 - 4 | 323 | 77 | |
XM_012964911.2 | XP_012820365.1 | 4 - 5 | 591 | 77 | ||
GenBank mRNA | BC121985.1 | - | 3 - 4 | 323 | 77 | |
BC135387.1 | - | 3 - 4 | 376 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |