Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079076.1 | NP_001072544.1 | 1 - 2 | 531 | 93 | |
GenBank mRNA | BC122048.1 | - | 1 - 2 | 531 | 93 | |
BC157219.1 | - | 1 - 2 | 529 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |