Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079127.1 | NP_001072595.1 | 2 - 3 | 1243 | 104 | |
XM_012965579.2 | XP_012821033.1 | 3 - 4 | 1084 | 104 | ||
XM_012965582.2 | XP_012821036.1 | 3 - 4 | 1007 | 104 | ||
XM_012965585.1 | XP_012821039.1 | 3 - 4 | 811 | 104 | ||
GenBank mRNA | BC122963.1 | - | 2 - 3 | 1243 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |