Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c11orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079316.1 | NP_001072784.1 | 1 - 2 | 58 | 80 | |
GenBank mRNA | BC118860.1 | - | 1 - 2 | 58 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |