Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079433.1 | NP_001072901.1 | 5 - 6 | 517 | 79 | |
GenBank mRNA | BC121430.1 | - | 5 - 6 | 517 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |