Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079569.1 | NP_001073037.1 | 3 - 4 | 1364 | 101 | |
XM_012963098.2 | XP_012818552.1 | 3 - 4 | 1439 | 101 | ||
XM_012963099.2 | XP_012818553.1 | 3 - 4 | 3002 | 101 | ||
GenBank mRNA | BC121242.1 | - | 3 - 4 | 1364 | 101 | |
CR942337.2 | - | 1 - 2 | 379 | 101 | ||
CT030382.1 | - | 3 - 4 | 1329 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |