Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083350.2 | NP_001076819.1 | 4 - 5 | 626 | 71 | |
XM_012958947.2 | XP_012814401.1 | 4 - 5 | 780 | 71 | ||
XM_018092059.1 | XP_017947548.1 | 3 - 4 | 655 | 71 | ||
XM_018092060.1 | XP_017947549.1 | 4 - 5 | 470 | 71 | ||
GenBank mRNA | BC088770.1 | - | 4 - 5 | 565 | 71 | |
BC135938.1 | - | 4 - 5 | 578 | 71 | ||
DN093113.1 | - | 4 - 5 | 626 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |