Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cracr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097181.1 | NP_001090650.1 | 1 - 2 | 332 | 115 | |
GenBank mRNA | BC127278.1 | - | 1 - 2 | 332 | 115 | |
CR761002.2 | - | 2 - 3 | 269 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |