Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ogfrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097287.1 | NP_001090756.1 | 1 - 2 | 290 | 102 | |
XM_012962530.2 | XP_012817984.1 | 1 - 2 | 593 | 102 | ||
GenBank mRNA | BC128651.1 | - | 1 - 2 | 290 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |