Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp3a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097314.1 | NP_001090783.1 | 8 - 9 | 782 | 88 | |
XM_012964140.2 | XP_012819594.1 | 7 - 8 | 785 | 88 | ||
XM_012964141.2 | XP_012819595.1 | 8 - 9 | 842 | 88 | ||
GenBank mRNA | BC135164.1 | - | 8 - 9 | 782 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |