Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sall2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004910746.3 | XP_004910803.1 | 1 - 2 | 520 | 99 | |
XM_004910750.3 | XP_004910807.1 | 1 - 2 | 518 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |