Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097378.1 | NP_001090847.1 | 7 - 8 | 1003 | 75 | |
GenBank mRNA | BC123095.1 | - | 7 - 8 | 1003 | 75 | |
BC158917.1 | - | 7 - 8 | 966 | 75 | ||
CU025106.1 | - | 7 - 8 | 1033 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |