Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dipk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097382.1 | NP_001090851.1 | 5 - 6 | 569 | 91 | |
XM_012960387.2 | XP_012815841.1 | 6 - 7 | 905 | 91 | ||
XM_012960388.2 | XP_012815842.1 | 5 - 6 | 846 | 91 | ||
GenBank mRNA | BC135136.1 | - | 5 - 6 | 569 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |