Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097397.1 | NP_001090866.1 | 6 - 7 | 1020 | 93 | |
XM_012956239.2 | XP_012811693.1 | 7 - 8 | 1135 | 93 | ||
GenBank mRNA | BC135300.1 | - | 6 - 7 | 1020 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |