Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vsx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100200.1 | NP_001093670.1 | 1 - 2 | 433 | 78 | |
XM_012962573.2 | XP_012818027.1 | 1 - 2 | 479 | 78 | ||
GenBank mRNA | BC135613.1 | - | 1 - 2 | 433 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |