Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100244.1 | NP_001093714.1 | 3 - 4 | 987 | 65 | |
GenBank mRNA | BC135743.1 | - | 3 - 4 | 987 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |