Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100247.1 | NP_001093717.1 | 4 - 5 | 1452 | 86 | |
GenBank mRNA | BC134803.1 | - | 4 - 5 | 1452 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |