Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: onecut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100260.1 | NP_001093730.1 | 2 - 3 | 1406 | 72 | |
GenBank mRNA | BC135775.2 | - | 2 - 3 | 1406 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |