Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfrsf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012965431.2 | XP_012820885.1 | 3 - 4 | 862 | 109 | |
GenBank mRNA | BC136133.1 | - | 3 - 4 | 383 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |