Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hdhd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100283.1 | NP_001093753.1 | 583 | 75 | ||
GenBank mRNA | BC136143.1 | - | 583 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |