Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bsx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101810.1 | NP_001095280.1 | 1 - 2 | 332 | 85 | |
GenBank mRNA | BC136001.1 | - | 1 - 2 | 332 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |