Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab11fip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC135580.1 | - | 3 - 4 | 1608 | 69 | |
CU075604.1 | - | 3 - 4 | 1537 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |