Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc39a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102697.1 | NP_001096167.1 | 475 | 105 | ||
GenBank mRNA | BC135963.1 | - | 475 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |