Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102721.1 | NP_001096191.1 | 3 - 4 | 399 | 77 | |
GenBank mRNA | BC136177.1 | - | 3 - 4 | 399 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |