Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csgalnact2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102722.1 | NP_001096192.1 | 3 - 4 | 1238 | 92 | |
XM_012965870.2 | XP_012821324.1 | 3 - 4 | 1308 | 92 | ||
XM_012965871.2 | XP_012821325.1 | 5 - 6 | 1463 | 92 | ||
XM_012965872.2 | XP_012821326.1 | 4 - 5 | 1409 | 92 | ||
XM_012965874.2 | XP_012821328.1 | 4 - 5 | 1362 | 92 | ||
XM_018095219.1 | XP_017950708.1 | 6 - 7 | 1577 | 92 | ||
XM_018095220.1 | XP_017950709.1 | 4 - 5 | 1452 | 92 | ||
GenBank mRNA | BC135663.1 | - | 3 - 4 | 1238 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |