Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102727.1 | NP_001096197.1 | 2 - 3 | 2245 | 91 | |
XM_012971500.2 | XP_012826954.1 | 2 - 3 | 2291 | 91 | ||
GenBank mRNA | BC135681.1 | - | 2 - 3 | 2245 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |