Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zc3h6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102732.2 | NP_001096202.2 | 9 - 10 | 1344 | 90 | |
GenBank mRNA | BC135562.1 | - | 9 - 10 | 1336 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |