Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102749.1 | NP_001096219.1 | 5 - 6 | 804 | 80 | |
GenBank mRNA | BC158230.1 | - | 5 - 6 | 802 | 80 | |
CR762165.2 | - | 5 - 6 | 804 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |